Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXIVsPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144485.3 | NP_001137957.1 | 2 - 3 | 381 | 87 | |
NM_001274974.1 | NP_001261903.1 | 2 - 3 | 376 | 87 | ||
NM_140558.4 | NP_648815.1 | 2 - 3 | 376 | 87 | ||
GenBank mRNA | AF349541.1 | - | 2 - 3 | 294 | 87 | |
AY094681.1 | - | 2 - 3 | 302 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |