Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104143.3 | NP_001097613.1 | 2 - 3 | 396 | 111 | |
NM_001382058.1 | NP_001369056.1 | 2 - 3 | 396 | 111 | ||
NM_140573.6 | NP_648830.1 | 2 - 3 | 396 | 111 | ||
GenBank mRNA | AY051508.1 | - | 2 - 3 | 362 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |