Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baldspot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140652.4 | NP_648909.1 | 2 - 3 | 380 | 76 | |
NM_168678.3 | NP_730187.2 | 2 - 3 | 440 | 76 | ||
GenBank mRNA | AF265296.1 | - | 2 - 3 | 392 | 76 | |
AY061121.1 | - | 2 - 3 | 392 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |