Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140719.4 | NP_648976.2 | 2 - 3 | 1001 | 124 | |
GenBank mRNA | BT150404.1 | - | 2 - 3 | 978 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |