Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140735.3 | NP_648992.1 | 1 - 2 | 949 | 59 | |
GenBank mRNA | BT099563.1 | - | 1 - 2 | 927 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |