Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275102.1 | NP_001262031.1 | 1 - 2 | 1107 | 55 | |
NM_001275103.1 | NP_001262032.1 | 1 - 2 | 1107 | 55 | ||
NM_140830.2 | NP_649087.1 | 1 - 2 | 1107 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |