Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr76Bc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104174.2 | NP_001097644.1 | 3 | 1335 | 61 | |
NM_001275122.1 | NP_001262051.1 | 3 | 1657 | 61 | ||
GenBank mRNA | BT022236.1 | - | 3 | 1445 | 61 | |
BT022288.1 | - | 3 | 1445 | 61 | ||
BT022352.1 | - | 3 | 1445 | 61 | ||
BT022412.1 | - | 3 | 1445 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |