Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144509.3 | NP_001137981.1 | 7 - 8 | 2176 | 58 | |
NM_001275130.2 | NP_001262059.1 | 7 - 8 | 2146 | 58 | ||
NM_001275131.1 | NP_001262060.1 | 7 - 8 | 1991 | 58 | ||
NM_140883.2 | NP_649140.1 | 7 - 8 | 2176 | 58 | ||
GenBank mRNA | BT133456.1 | - | 7 - 8 | 2175 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |