Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGDI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275151.1 | NP_001262080.1 | 2 - 3 | 270 | 72 | |
NM_140905.3 | NP_649162.1 | 2 - 3 | 368 | 72 | ||
GenBank mRNA | AY061221.1 | - | 2 - 3 | 304 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |