Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toll-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259916.2 | NP_001246845.1 | 1 - 2 | 298 | 72 | |
NM_001259917.1 | NP_001246846.1 | 1 - 2 | 298 | 72 | ||
NM_140957.4 | NP_649214.1 | 2 - 3 | 883 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |