Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toll-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259916.2 | NP_001246845.1 | 4 - 5 | 2282 | 115 | |
NM_001259917.1 | NP_001246846.1 | 5 - 6 | 2504 | 115 | ||
NM_140957.4 | NP_649214.1 | 6 - 7 | 3089 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |