Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273474.1 | NP_001260403.1 | 2 - 3 | 1059 | 67 | |
NM_165002.2 | NP_723737.1 | 2 - 3 | 1206 | 67 | ||
GenBank mRNA | BT133212.1 | - | 2 - 3 | 1185 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |