Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169492.2 | NP_001162963.1 | 4 - 5 | 1047 | 67 | |
NM_001273474.1 | NP_001260403.1 | 5 - 6 | 1685 | 67 | ||
NM_001273475.1 | NP_001260404.1 | 4 - 5 | 919 | 67 | ||
NM_001273476.1 | NP_001260405.1 | 4 - 5 | 772 | 67 | ||
NM_001273477.1 | NP_001260406.1 | 4 - 5 | 1077 | 67 | ||
NM_001273478.1 | NP_001260407.1 | 4 - 5 | 1048 | 67 | ||
NM_165002.2 | NP_723737.1 | 5 - 6 | 1832 | 67 | ||
NM_165003.3 | NP_723738.2 | 4 - 5 | 1047 | 67 | ||
NM_165004.3 | NP_723739.1 | 4 - 5 | 1047 | 67 | ||
GenBank mRNA | AY051758.1 | - | 4 - 5 | 1027 | 67 | |
BT133212.1 | - | 5 - 6 | 1811 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |