Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165020.3 | NP_723772.2 | 4 - 5 | 2013 | 89 | |
NM_205972.2 | NP_995694.1 | 4 - 5 | 2009 | 89 | ||
NM_205973.2 | NP_995695.1 | 5 - 6 | 2127 | 89 | ||
NM_205974.2 | NP_995696.1 | 4 - 5 | 2009 | 89 | ||
NM_205975.2 | NP_995697.1 | 4 - 5 | 2009 | 89 | ||
NM_205976.2 | NP_995698.1 | 4 - 5 | 2009 | 89 | ||
NM_205977.2 | NP_995699.1 | 5 - 6 | 2127 | 89 | ||
NM_205979.2 | NP_995701.1 | 5 - 6 | 2127 | 89 | ||
NM_205980.2 | NP_995702.1 | 5 - 6 | 2127 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |