Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yuri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316397.1 | NP_001303326.1 | 6 - 7 | 1557 | 70 | |
NM_165106.5 | NP_723903.3 | 5 - 6 | 1435 | 70 | ||
NM_165107.3 | NP_723904.1 | 5 - 6 | 1900 | 70 | ||
NM_165109.3 | NP_723906.1 | 6 - 7 | 1845 | 70 | ||
GenBank mRNA | BT001564.1 | - | 5 - 6 | 1898 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |