Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yuri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038821.2 | NP_001033910.2 | 3 - 4 | 749 | 66 | |
NM_001038822.3 | NP_001033911.3 | 4 - 5 | 663 | 66 | ||
NM_001316397.1 | NP_001303326.1 | 7 - 8 | 1657 | 66 | ||
NM_165106.5 | NP_723903.3 | 6 - 7 | 1535 | 66 | ||
NM_165107.3 | NP_723904.1 | 6 - 7 | 2000 | 66 | ||
NM_165109.3 | NP_723906.1 | 7 - 8 | 1945 | 66 | ||
GenBank mRNA | BT001564.1 | - | 6 - 7 | 1998 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |