Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299053.1 | NP_001285982.1 | 1 - 2 | 248 | 89 | |
NM_057908.4 | NP_477256.1 | 1 - 2 | 248 | 89 | ||
NM_165157.2 | NP_723966.1 | 1 - 2 | 248 | 89 | ||
NM_165158.1 | NP_723967.1 | 2 - 3 | 323 | 89 | ||
GenBank mRNA | AY047561.1 | - | 1 - 2 | 229 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |