Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300164.1 | NP_001287093.1 | 2 - 3 | 278 | 58 | |
NM_170638.2 | NP_730265.1 | 1 - 2 | 246 | 58 | ||
GenBank mRNA | BT133104.1 | - | 2 - 3 | 281 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |