Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shakB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103568.3 | NP_001097038.2 | 4 - 5 | 1849 | 113 | |
NM_001144750.2 | NP_001138222.1 | 3 - 4 | 590 | 113 | ||
NM_001258858.2 | NP_001245787.1 | 7 - 8 | 1701 | 113 | ||
NM_001258859.2 | NP_001245788.1 | 8 - 9 | 2565 | 113 | ||
NM_001298574.1 | NP_001285503.1 | 7 - 8 | 2806 | 113 | ||
NM_078701.3 | NP_523425.2 | 6 - 7 | 1997 | 113 | ||
NM_134566.3 | NP_608410.2 | 5 - 6 | 1101 | 113 | ||
NM_170660.2 | NP_728361.1 | 3 - 4 | 1349 | 113 | ||
GenBank mRNA | S78495.1 | - | 3 - 4 | 1349 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |