Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NfI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272122.2 | NP_001259051.1 | 2 - 3 | 218 | 80 | |
NM_205870.3 | NP_995592.2 | 2 - 3 | 218 | 80 | ||
GenBank mRNA | BT003547.1 | - | 2 - 3 | 216 | 80 | |
BT023941.1 | - | 2 - 3 | 216 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |