Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258491.3 | NP_001245420.1 | 2 - 3 | 844 | 81 | |
NM_079882.4 | NP_524621.2 | 2 - 3 | 770 | 81 | ||
NM_166756.3 | NP_726566.1 | 2 - 3 | 466 | 81 | ||
NM_166757.3 | NP_726567.2 | 2 - 3 | 770 | 81 | ||
NM_205873.3 | NP_995595.1 | 2 - 3 | 466 | 81 | ||
GenBank mRNA | AJ320236.1 | - | 1 - 2 | 423 | 81 | |
BT015268.1 | - | 2 - 3 | 731 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |