Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: for | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014464.2 | NP_001014464.1 | 2 - 3 | 1823 | 58 | |
NM_001169387.2 | NP_001162858.1 | 3 - 4 | 1703 | 58 | ||
NM_001347740.1 | NP_001334731.1 | 3 - 4 | 1373 | 58 | ||
NM_001369953.1 | NP_001356892.1 | 4 - 5 | 953 | 58 | ||
NM_001369954.1 | NP_001356896.1 | 3 - 4 | 674 | 58 | ||
NM_001369955.1 | NP_001356955.1 | 2 - 3 | 404 | 58 | ||
NM_058139.4 | NP_477487.1 | 5 - 6 | 2532 | 58 | ||
NM_058141.4 | NP_477489.1 | 3 - 4 | 1715 | 58 | ||
NM_058142.4 | NP_477490.1 | 2 - 3 | 2275 | 58 | ||
NM_134319.4 | NP_599146.1 | 3 - 4 | 2155 | 58 | ||
NM_205904.2 | NP_995626.1 | 5 - 6 | 2518 | 58 | ||
NM_205906.2 | NP_995628.1 | 4 - 5 | 2253 | 58 | ||
NM_205907.2 | NP_995629.1 | 3 - 4 | 1763 | 58 | ||
GenBank mRNA | AF132175.2 | - | 3 - 4 | 1686 | 58 | |
AY061514.1 | - | 5 - 6 | 2517 | 58 | ||
BT004881.1 | - | 3 - 4 | 2191 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |