Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169404.2 | NP_001162875.1 | 4 - 5 | 1988 | 83 | |
NM_001169405.2 | NP_001162876.1 | 5 - 6 | 747 | 83 | ||
NM_001169406.2 | NP_001162877.1 | 5 - 6 | 730 | 83 | ||
NM_001169407.2 | NP_001162878.2 | 3 - 4 | 727 | 83 | ||
NM_175969.3 | NP_787983.1 | 3 - 4 | 1952 | 83 | ||
NM_175970.3 | NP_787984.1 | 5 - 6 | 791 | 83 | ||
NM_175971.3 | NP_787985.1 | 4 - 5 | 763 | 83 | ||
NM_175972.3 | NP_787986.1 | 4 - 5 | 695 | 83 | ||
NM_175973.3 | NP_787987.1 | 3 - 4 | 680 | 83 | ||
NM_175974.3 | NP_787988.1 | 3 - 4 | 519 | 83 | ||
NM_205908.2 | NP_995630.1 | 3 - 4 | 690 | 83 | ||
GenBank mRNA | AY069432.1 | - | 5 - 6 | 757 | 83 | |
BT099515.1 | - | 3 - 4 | 507 | 83 | ||
BT099529.1 | - | 3 - 4 | 676 | 83 | ||
BT132825.1 | - | 4 - 5 | 665 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |