Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298825.1 | NP_001285754.1 | 5 - 6 | 2793 | 75 | |
NM_135386.5 | NP_609230.2 | 5 - 6 | 2782 | 75 | ||
NM_205938.2 | NP_995660.2 | 5 - 6 | 2782 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |