Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014474.4 | NP_001014474.3 | 4 - 5 | 921 | 87 | |
NM_001169424.2 | NP_001162895.2 | 4 - 5 | 921 | 87 | ||
NM_001298737.1 | NP_001285666.1 | 4 - 5 | 921 | 87 | ||
GenBank mRNA | BT128907.1 | - | 4 - 5 | 920 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |