Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298739.1 | NP_001285668.1 | 1 - 2 | 1789 | 63 | |
NM_135176.3 | NP_609020.3 | 2 - 3 | 2202 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |