Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298746.1 | NP_001285675.1 | 1 - 2 | 326 | 54 | |
NM_135191.3 | NP_609035.2 | 1 - 2 | 326 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |