Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spn28Dc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135328.3 | NP_609172.1 | 1 - 2 | 779 | 64 | |
GenBank mRNA | BT031146.1 | - | 1 - 2 | 822 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |