Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 22 - 23 | 6019 | 67 | |
NM_001169436.3 | NP_001162907.1 | 22 - 23 | 6064 | 67 | ||
NM_001316385.1 | NP_001303314.1 | 23 - 24 | 6925 | 67 | ||
NM_135344.6 | NP_609188.4 | 22 - 23 | 5808 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |