Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273361.1 | NP_001260290.1 | 3 - 4 | 574 | 72 | |
NM_135441.2 | NP_609285.1 | 3 - 4 | 744 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |