Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273397.1 | NP_001260326.1 | 3 - 4 | 2087 | 74 | |
NM_001273398.1 | NP_001260327.1 | 3 - 4 | 1907 | 74 | ||
NM_135539.3 | NP_609383.2 | 3 - 4 | 1907 | 74 | ||
GenBank mRNA | BT133312.1 | - | 3 - 4 | 1899 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |