Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273404.1 | NP_001260333.1 | 1 - 2 | 246 | 71 | |
NM_135545.3 | NP_609389.1 | 1 - 2 | 246 | 71 | ||
GenBank mRNA | BT133409.1 | - | 1 - 2 | 115 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |