Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273404.1 | NP_001260333.1 | 3 - 4 | 922 | 89 | |
NM_135545.3 | NP_609389.1 | 3 - 4 | 922 | 89 | ||
GenBank mRNA | BT133409.1 | - | 3 - 4 | 791 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |