Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273407.2 | NP_001260336.1 | 3 - 4 | 671 | 65 | |
NM_001273408.2 | NP_001260337.1 | 4 - 5 | 480 | 65 | ||
NM_135548.3 | NP_609392.1 | 3 - 4 | 692 | 65 | ||
GenBank mRNA | AY122217.1 | - | 3 - 4 | 678 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |