Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273407.2 | NP_001260336.1 | 6 - 7 | 1364 | 55 | |
NM_001273408.2 | NP_001260337.1 | 7 - 8 | 1173 | 55 | ||
NM_135548.3 | NP_609392.1 | 6 - 7 | 1385 | 55 | ||
GenBank mRNA | AY122217.1 | - | 6 - 7 | 1371 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |