Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nse4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169467.1 | NP_001162938.1 | 4 - 5 | 895 | 69 | |
NM_135561.3 | NP_609405.2 | 4 - 5 | 895 | 69 | ||
GenBank mRNA | BT023592.1 | - | 4 - 5 | 895 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |