Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr36c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165207.1 | NP_724040.1 | 2 - 3 | 1023 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Drosophila melanogaster |