Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG31750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165208.1 | NP_724041.1 | 1 - 2 | 658 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |