Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103703.2 | NP_001097173.1 | 2 - 3 | 942 | 67 | |
NM_001103704.3 | NP_001097174.1 | 2 - 3 | 942 | 67 | ||
NM_001169534.3 | NP_001163005.1 | 2 - 3 | 942 | 67 | ||
NM_001299109.1 | NP_001286038.1 | 2 - 3 | 942 | 67 | ||
NM_001299110.1 | NP_001286039.1 | 2 - 3 | 942 | 67 | ||
NM_165249.4 | NP_724106.1 | 2 - 3 | 942 | 67 | ||
NM_165250.4 | NP_724107.1 | 2 - 3 | 942 | 67 | ||
GenBank mRNA | AY075516.1 | - | 2 - 3 | 937 | 67 | |
BT133041.1 | - | 2 - 3 | 921 | 67 | ||
M27813.1 | - | 2 - 3 | 940 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |