Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mst36Fa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165259.3 | NP_724121.3 | 1 - 2 | 768 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |