Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ham | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259152.1 | NP_001246081.1 | 7 - 8 | 2470 | 68 | |
NM_001273614.1 | NP_001260543.1 | 7 - 8 | 2802 | 68 | ||
NM_001273615.1 | NP_001260544.1 | 4 - 5 | 1435 | 68 | ||
NM_001273616.1 | NP_001260545.2 | 6 - 7 | 2339 | 68 | ||
NM_165262.3 | NP_724130.4 | 7 - 8 | 2471 | 68 | ||
GenBank mRNA | AY166585.1 | - | 6 - 7 | 1821 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |