Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259156.1 | NP_001246085.1 | 2 - 3 | 842 | 61 | |
NM_001259157.3 | NP_001246086.1 | 1 - 2 | 553 | 61 | ||
NM_001259159.1 | NP_001246088.1 | 2 - 3 | 1484 | 61 | ||
NM_001273630.2 | NP_001260559.1 | 1 - 2 | 553 | 61 | ||
NM_165277.3 | NP_724161.1 | 2 - 3 | 518 | 61 | ||
GenBank mRNA | BT029915.1 | - | 2 - 3 | 515 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |