Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169549.1 | NP_001163020.1 | 10 - 11 | 2147 | 73 | |
NM_001259172.2 | NP_001246101.1 | 10 - 11 | 2144 | 73 | ||
NM_001259173.1 | NP_001246102.1 | 10 - 11 | 2150 | 73 | ||
NM_001299171.1 | NP_001286100.1 | 10 - 11 | 2150 | 73 | ||
NM_080115.3 | NP_524854.2 | 10 - 11 | 2150 | 73 | ||
NM_165323.3 | NP_724254.1 | 10 - 11 | 2150 | 73 | ||
NM_165324.2 | NP_724255.1 | 10 - 11 | 2147 | 73 | ||
NM_165325.3 | NP_724256.2 | 2 - 3 | 706 | 73 | ||
GenBank mRNA | AJ238876.1 | - | 10 - 11 | 1797 | 73 | |
AY075586.1 | - | 10 - 11 | 2136 | 73 | ||
BT044175.1 | - | 2 - 3 | 681 | 73 | ||
BT122081.1 | - | 10 - 11 | 1826 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |