Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKKepsilon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136204.3 | NP_610048.2 | 4 - 5 | 1565 | 134 | |
NM_165337.2 | NP_724278.1 | 3 - 4 | 1475 | 134 | ||
GenBank mRNA | BT046140.2 | - | 3 - 4 | 1449 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |