Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165372.1 | NP_724331.1 | 1 - 2 | 907 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |