Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165375.3 | NP_724335.2 | 4 - 5 | 917 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |