Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165376.1 | NP_724336.1 | 1 - 2 | 882 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Drosophila melanogaster |