Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273746.2 | NP_001260675.1 | 8 - 9 | 1370 | 72 | |
NM_136264.6 | NP_610108.1 | 7 - 8 | 1224 | 72 | ||
NM_165378.1 | NP_724339.1 | 7 - 8 | 1380 | 72 | ||
NM_165379.3 | NP_724340.1 | 7 - 8 | 1247 | 72 | ||
GenBank mRNA | AF202633.1 | - | 7 - 8 | 1144 | 72 | |
AY314744.1 | - | 7 - 8 | 1151 | 72 | ||
BT044501.1 | - | 7 - 8 | 1380 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |