Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136336.3 | NP_610180.1 | 3 - 4 | 602 | 93 | |
NM_165441.2 | NP_724424.1 | 3 - 4 | 949 | 93 | ||
GenBank mRNA | AY047548.1 | - | 3 - 4 | 574 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |