Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103732.2 | NP_001097202.2 | 16 - 17 | 2876 | 100 | |
NM_001273807.1 | NP_001260736.1 | 13 - 14 | 2520 | 100 | ||
NM_001273808.1 | NP_001260737.2 | 12 - 13 | 2139 | 100 | ||
NM_165490.2 | NP_724498.1 | 13 - 14 | 2521 | 100 | ||
GenBank mRNA | AY058255.1 | - | 13 - 14 | 2504 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |